Sterilin™ Water Sampling Bottles
Supplier: LAB 3 BOTTLES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WSC350/NT.1 WSC500-20.1 WSC1000-20.1 WSC1000/NT.1 WSC500/NT.1 WSC1000.1 WSC350.1 WSC500.1 WSC500T.1 WSC500T/NT.1 WSC500T-20.1 WSC350-20.1
PLBOWSC350/NT.1EA 0 GBP
PLBOWSC350/NT.1 PLBOWSC500-20.1 PLBOWSC1000-20.1 PLBOWSC1000/NT.1 PLBOWSC500/NT.1 PLBOWSC1000.1 PLBOWSC350.1 PLBOWSC500.1 PLBOWSC500T.1 PLBOWSC500T/NT.1 PLBOWSC500T-20.1 PLBOWSC350-20.1
Sterilin™ Water Sampling Bottles
Bottles Water Sampling Bottles
Conveniently sample both chlorinated or nonchlorinated water for microbial analysis with Sterilin™ water sampling bottles.
  • Tamper evident for sample integrity
  • Lot number for complete traceability
  • Sterilized by gamma irradiation
  • Available empty, or pre-dosed with sodium thiosulphate
  • Flexible polystyrene
  • NEW, lightweight PET for improved handling
  • Closure is made up of HDPE
Polystyrene bottles feature color-coded labels to aid differentiation: Blue (dosed) and green (undosed).
Available dose levels:
20 mg/h: Suitable for neutralising water samples with a low concentration of chlorine
120 mg/h: Suitable for neutralising water samples with a high concentration of chlorine