Octadecyltrichlorosilane
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Trichloro(stearyl)silane, Trichlorooctadecylsilane, Octadecyltrichlorosilane, ODTS, Trichloro(octadecyl)silane
SILS12250.25
SILS12250.100
SILS12250.250
SILNSILS12250.25EA
0
GBP
SILNSILS12250.25
SILNSILS12250.100
SILNSILS12250.250
Octadecyltrichlorosilane
Octadecyltrichlorosilane
Formula:
C₁₈H₃₇Cl₃Si MW: 387.94 g/mol Boiling Pt: 361 °C (1013 hPa) Melting Pt: 22 °C Density: 0.98 g/cm³ (20 °C) Flash Pt: 89 °C |
MDL Number:
MFCD00000484 CAS Number: 112-04-9 EINECS: 203-930-7 UN: 1800 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...