2,4,5-Trimethylthiazole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ300

W332518-100G-K W332518-1KG-K W332518-25G-K
SIALW332518-100G-KEA 229 GBP
SIALW332518-100G-K SIALW332518-1KG-K SIALW332518-25G-K
2,4,5-Trimethylthiazole, Sigma-Aldrich®
2,4,5-Trimethylthiazole
Formula: C₆H₉NS
Boiling Pt: 165 °C (1013 hPa)
Density: 1.013 g/cm³ (16 °C)
MDL Number: MFCD00005332
CAS Number: 13623-11-5
EINECS: 237-107-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR