trans-3-Hexenoic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ300

W317004-100G-K W317004-1KG-K
SIALW317004-100G-KEA 145 GBP
SIALW317004-100G-K SIALW317004-1KG-K
trans-3-Hexenoic acid, Sigma-Aldrich®
trans-3-Hexenoic acid
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 209 °C (1013 hPa)
Melting Pt: 14 °C
Density: 0.965 g/cm³ (20 °C)
MDL Number: MFCD00002786
CAS Number: 1577-18-0
EINECS: 216-417-8
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR