trans-2-Hexenoic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
trans-Hex-2-enoic acid
M-Clarity™ quality level = MQ300
W316903-100G-K
W316903-1KG-K
W316903-5KG-K
SIALW316903-100G-KEA
146
GBP
SIALW316903-100G-K
SIALW316903-1KG-K
SIALW316903-5KG-K
trans-2-Hexenoic acid, Sigma-Aldrich®
trans-2-Hexenoic acid
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 217 °C (1013 hPa) Melting Pt: 32 °C Density: 0.946 g/cm³ (45 °C) |
MDL Number:
MFCD00002705 CAS Number: 13419-69-7 EINECS: 236-528-5 UN: 3261 ADR: 8,III |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...