Propyl propionate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ100

W295809-1KG-K W295809-4KG-K W295809-9KG-K
SIALW295809-1KG-KEA 83.8 GBP
SIALW295809-1KG-K SIALW295809-4KG-K SIALW295809-9KG-K
Propyl propionate, Sigma-Aldrich®
Formula: C₆H₁₂O₂
Density: 0.88 g/cm³ (20 °C)
MDL Number: MFCD00009373
CAS Number: 106-36-5
EINECS: 203-389-7
UN: 3272
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR