(±)-γ-Valerolactone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(±)-4-Hydroxypentanoic acid lactone, (±)-Pentan-4-olide, γ-Pentanolactone, 5-Methyldihydrofuran-2(3H)-one, gamma-Valerolactone
M-Clarity™ quality level = MQ100
V403-100G
V403-500G
SIALV403-100GEA
111
GBP
SIALV403-100G
SIALV403-500G
(±)-γ-Valerolactone, Sigma-Aldrich®
(±)-γ-Valerolactone
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 207 °C (1013 hPa) Melting Pt: –31 °C Density: 1.05 g/cm³ (20 °C) |
MDL Number:
MFCD00005400 CAS Number: 108-29-2 EINECS: 203-569-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...