D-(-)-Ribose, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

R7500-100G R7500-10MG R7500-25G R7500-5G
SIALR7500-100GEA 300 GBP
SIALR7500-100G SIALR7500-10MG SIALR7500-25G SIALR7500-5G
D-(-)-Ribose, Sigma-Aldrich®
D(-)-Ribose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 90…95 °C
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135453
CAS Number: 50-69-1
EINECS: 200-059-4
Merck Index: 12,08371

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR