1-Phenyl-2-thiourea, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
N-Phenylthiourea, 1-Phenylthiourea
P7629-100G
P7629-10G
P7629-25G
SIALP7629-100GEA
266
GBP
SIALP7629-100G
SIALP7629-10G
SIALP7629-25G
1-Phenyl-2-thiourea, Sigma-Aldrich®
1-Phenyl-2-thiourea
Formula:
C₇H₈N₂S MW: 152.22 g/mol Boiling Pt: 295…297 °C (1013 hPa) Melting Pt: 84…85 °C Density: 1.33 g/cm³ (20 °C) |
MDL Number:
MFCD00004933 CAS Number: 103-85-5 EINECS: 203-151-2 UN: 2811 ADR: 6.1,I Merck Index: 12,07468 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...