Ethyl chloroacetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ200

E16856-1L E16856-2.5L E16856-250ML
SIALE16856-1LEA 104 GBP
SIALE16856-1L SIALE16856-2.5L SIALE16856-250ML
Ethyl chloroacetate, Sigma-Aldrich®
Ethyl chloroacetate
Formula: C₄H₇ClO₂
MW: 122.55 g/mol
Boiling Pt: 144
Melting Pt: –26 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 56
MDL Number: MFCD00000932
CAS Number: 105-39-5
EINECS: 203-294-0
UN: 1181
ADR: 6.1 II
Merck Index: 13,03818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR