2,5-Diphenyloxazole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ200

D4630-100G D4630-1KG D4630-500G
SIALD4630-100GEA 146 GBP
SIALD4630-100G SIALD4630-1KG SIALD4630-500G
2,5-Diphenyloxazole, Sigma-Aldrich®
2,5-Diphenyloxazole
Formula: C₁₅H₁₁NO
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 70…72 °C
Density: 1.094 g/cm³ (99 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005306
CAS Number: 92-71-7
EINECS: 202-181-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR