Cottonseed oil, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

C7767-1L C7767-500ML
SIALC7767-1LEA 89.5 GBP
SIALC7767-1L SIALC7767-500ML
Cottonseed oil, Sigma-Aldrich®
Cottonseed oil
Density: 0.91 g/cm³ (20 °C)
MDL Number: MFCD00130872
CAS Number: 8001-29-4
EINECS: 232-280-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR