2-Amino-4,6-dihydroxypyrimidine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Amino-6-hydroxy-1H-pyrimidin-4-one, 2-Amino-6-hydroxypyrimidin-4(3H)-one
M-Clarity™ quality level = MQ100
A50401-100G
A50401-25G
SIALA50401-100GEA
762
GBP
SIALA50401-100G
SIALA50401-25G
2-Amino-4,6-dihydroxypyrimidine, Sigma-Aldrich®
2-Amino-4,6-dihydroxypyrimidine
Formula:
C₄H₅N₃O₂ MW: 127.1 g/mol Boiling Pt: 394 °C (1013 hPa) Melting Pt: 300 °C |
MDL Number:
MFCD00006094 CAS Number: 56-09-7 EINECS: 200-256-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...