3-Amino-2-(trifluoromethyl)pyridine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(Trifluoromethyl)-3-pyridylamine, 2-(Trifluoromethyl)pyridin-3-amine
M-Clarity™ quality level = MQ100
686972-1G
686972-250MG
SIAL686972-1GEA
353
GBP
SIAL686972-1G
SIAL686972-250MG
3-Amino-2-(trifluoromethyl)pyridine, Sigma-Aldrich®
3-Amino-2-(trifluoromethyl)pyridine
Formula:
C₆H₅F₃N₂ MW: 162.11 g/mol |
MDL Number:
MFCD09753488 CAS Number: 106877-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...