2-Amino-4-(trifluoromethyl)pyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Trifluoromethyl)pyridin-2-amine, 4-(Trifluoromethyl)-2-pyridylamine

M-Clarity™ quality level = MQ200

660345-100MG 660345-500MG
SIAL660345-100MGEA 106 GBP
SIAL660345-100MG SIAL660345-500MG
2-Amino-4-(trifluoromethyl)pyridine, Sigma-Aldrich®
2-Amino-4-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
MDL Number: MFCD00077455
CAS Number: 106447-97-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR