N,N-Dimethylethylamine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: DMEA

M-Clarity™ quality level = MQ100

652571-2L 652571-500ML
SIAL652571-2LEA 568 GBP
SIAL652571-2L SIAL652571-500ML
N,N-Dimethylethylamine, Sigma-Aldrich®
N,N-Dimethylethylamine
Formula: C₄H₁₁N
Boiling Pt: 36 °C (1013 hPa)
Melting Pt: –140 °C
Density: 0.675 g/cm³ (20 °C)
Flash Pt: –45 °C
MDL Number: MFCD00009039
CAS Number: 598-56-1
EINECS: 209-940-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR