4-Fluorophthalic anhydride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Fluoroisobenzofuran-1,3-dione

M-Clarity™ quality level = MQ200

641375-25G 641375-5G
SIAL641375-25GEA 416 GBP
SIAL641375-25G SIAL641375-5G
4-Fluorophthalic anhydride, Sigma-Aldrich®
4-Fluorophthalic anhydride
Formula: C₈H₃FO₃
MW: 166.11 g/mol
Boiling Pt: 260
Melting Pt: 79…80 °C
Density: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00191363
CAS Number: 319-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR