2,6-Dichlorobenzothiazole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

553670-1G 553670-5G
SIAL553670-1GEA 37.2 GBP
SIAL553670-1G SIAL553670-5G
2,6-Dichlorobenzothiazole, Sigma-Aldrich®
2,6-Dichlorobenzothiazole
Formula: C₇H₃Cl₂NS
MW: 204.08 g/mol
MDL Number: MFCD00044101
CAS Number: 3622-23-9
EINECS: 222-819-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR