4-Phenylbenzylamine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

552313-1G 552313-5G
SIAL552313-1GEA 46.3 GBP
SIAL552313-1G SIAL552313-5G
4-Phenylbenzylamine, Sigma-Aldrich®
4-Phenylbenzylamine
Formula: C₁₃H₁₃N
MDL Number: MFCD01310838
CAS Number: 712-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR