4-Bromo-3,5-dimethylisoxazole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

544299-1G 544299-5G
SIAL544299-1GEA 37.2 GBP
SIAL544299-1G SIAL544299-5G
4-Bromo-3,5-dimethylisoxazole, Sigma-Aldrich®
4-Bromo-3,5-dimethylisoxazole
Formula: C₅H₆BrNO
MW: 176.01 g/mol
MDL Number: MFCD00068187
CAS Number: 10558-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR