2-Fluoro-6-(trifluoromethyl)benzyl bromide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(Bromomethyl)-2-fluoro-6-(trifluoromethyl)benzene, 2-(Bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene

M-Clarity™ quality level = MQ200

539627-1G 539627-5G
SIAL539627-1GEA 42.5 GBP
SIAL539627-1G SIAL539627-5G
2-Fluoro-6-(trifluoromethyl)benzyl bromide, Sigma-Aldrich®
2-Fluoro-6-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrF₄
MW: 257.03 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 43…45 °C
MDL Number: MFCD00082477
CAS Number: 239087-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR