3-Phenylbenzyl bromide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Bromomethyl)biphenyl, m-(Bromomethyl)biphenyl

M-Clarity™ quality level = MQ200

538388-25G 538388-5G
SIAL538388-25GEA 395 GBP
SIAL538388-25G SIAL538388-5G
3-Phenylbenzyl bromide, Sigma-Aldrich®
3-Phenylbenzyl bromide
Formula: C₁₃H₁₁Br
MW: 247.13 g/mol
MDL Number: MFCD03452790
CAS Number: 14704-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR