4-Cyanophenyl isocyanate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
493783-10G
493783-2G
SIAL493783-10GEA
626
GBP
SIAL493783-10G
SIAL493783-2G
4-Cyanophenyl isocyanate, Sigma-Aldrich®
4-Cyanophenyl isocyanate
Formula:
C₈H₄N₂O MW: 144.13 g/mol Boiling Pt: 123 °C (12 mmHg) Melting Pt: 99 °C |
CAS Number:
40465-45-0 EINECS: 254-934-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...