2-Phenylbenzyl bromide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: o-(Bromomethyl)biphenyl, 2-(Bromomethyl)biphenyl, 2-Phenylbenzylbromide

360082-25G 360082-5G
SIAL360082-25GEA 586 GBP
SIAL360082-25G SIAL360082-5G
2-Phenylbenzyl bromide, Sigma-Aldrich®
2-Phenylbenzyl bromide
Formula: C₁₃H₁₁Br
MW: 247.13 g/mol
MDL Number: MFCD00075454
CAS Number: 19853-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR