4-Nitrophthalic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
274755-25G
274755-5G
SIAL274755-25GEA
77.5
GBP
SIAL274755-25G
SIAL274755-5G
4-Nitrophthalic acid, Sigma-Aldrich®
4-Nitrophthalic acid
Formula:
C₈H₅NO₆ MW: 211.13 g/mol Melting Pt: 162…165 °C |
MDL Number:
MFCD00007252 CAS Number: 610-27-5 EINECS: 210-215-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...