4-tert-Butylbenzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-tert-Butylbenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ100

269670-10G 269670-50G
SIAL269670-10GEA 91 GBP
SIAL269670-10G SIAL269670-50G
4-tert-Butylbenzenesulphonyl chloride, Sigma-Aldrich®
4-tert-Butylbenzenesulphonyl chloride
Formula: C₁₀H₁₃ClO₂S
MW: 232.73 g/mol
Boiling Pt: 165 °C (18 mmHg)
Melting Pt: 83 °C
MDL Number: MFCD00007449
CAS Number: 15084-51-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR