Oil of castor, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi
M-Clarity™ quality level = MQ200
259853-250ML
259853-1L
SIAL259853-250MLEA
40.3
GBP
SIAL259853-250ML
SIAL259853-1L
Oil of castor, Sigma-Aldrich®
Oil of castor
Boiling Pt:
313 Melting Pt: –10 °C Density: 0.95 g/cm³ (20 °C) Flash Pt: >113 °C (closed cup) |
MDL Number:
MFCD00130746 CAS Number: 8001-79-4 EINECS: 232-293-8 Merck Index: 13,01908 |