(4-Chlorophenoxy)acetic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-CPA

25890-1KG 25890-250G
SIAL25890-1KGEA 266 GBP
SIAL25890-1KG SIAL25890-250G
(4-Chlorophenoxy)acetic acid, Sigma-Aldrich®
(4-Chlorophenoxy)acetic acid
Formula: (C₆H₄Cl)OCH₂COOH
MW: 186.59 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.53 g/cm³ (20 °C)
MDL Number: MFCD00004305
CAS Number: 122-88-3
EINECS: 204-581-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR