Methyldiphenylphosphine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Diphenylmethylphosphine
M-Clarity™ quality level = MQ100
244902-10G
244902-1G
244902-50G
SIAL244902-10GEA
155
GBP
SIAL244902-10G
SIAL244902-1G
SIAL244902-50G
Methyldiphenylphosphine, Sigma-Aldrich®
Methyldiphenylphosphine
Formula:
C₁₃H₁₃P MW: 200.22 g/mol Boiling Pt: 308 °C (1013 hPa) Melting Pt: 28 °C Density: 1.0784 g/cm³ (15 °C) |
MDL Number:
MFCD00008508 CAS Number: 1486-28-8 EINECS: 216-065-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...