2,2,2-Trichloroethyl chloroformate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
23258-100ML
23258-25ML
SIAL23258-100MLEA
318
GBP
SIAL23258-100ML
SIAL23258-25ML
2,2,2-Trichloroethyl chloroformate, Sigma-Aldrich®
2,2,2-Trichloroethyl chloroformate
Formula:
C₃H₂Cl₄O₂ Boiling Pt: 171 °C (1013 hPa) Melting Pt: 1 °C Density: 1.53 g/cm³ (20 °C) |
MDL Number:
MFCD00000810 CAS Number: 17341-93-4 EINECS: 241-363-7 UN: 3277 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...