Benzyltributylammonium chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: N-Benzyl-N,N-dibutylbutan-1-aminium chloride

M-Clarity™ quality level = MQ200

193771-100G 193771-500G
SIAL193771-100GEA 74.2 GBP
SIAL193771-100G SIAL193771-500G
Benzyltributylammonium chloride, Sigma-Aldrich®
Benzyltributylammonium chloride
Formula: C₁₉H₃₄ClN
MW: 311.94 g/mol
Melting Pt: 155…159 °C
MDL Number: MFCD00011849
CAS Number: 23616-79-7
EINECS: 245-787-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR