1-Butyl-3-methylimidazoliumhexafluorophosphate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
18122-50G-F
18122-5G-F
SIAL18122-50G-FEA
1040
GBP
SIAL18122-50G-F
SIAL18122-5G-F
1-Butyl-3-methylimidazoliumhexafluorophosphate, Sigma-Aldrich®
1-Butyl-3-methylimidazolium hexafluorophosphate
Formula:
C₈H₁₅F₆N₂P Boiling Pt: ∼200 °C (1013 hPa) Melting Pt: ∼6 °C Density: 1.38 g/cm³ (20 °C) Flash Pt: >110 °C |
MDL Number:
MFCD03093295 CAS Number: 174501-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...