α-Bromo-p-toluic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Bromomethylbenzoic acid, 4-(Bromomethyl)benzoic acid
M-Clarity™ quality level = MQ200
159549-10G
159549-50G
159549-5G
SIAL159549-10GEA
122
GBP
SIAL159549-10G
SIAL159549-50G
SIAL159549-5G
α-Bromo-p-toluic acid, Sigma-Aldrich®
α-Bromo-p-toluic acid
Formula:
C₈H₇BrO₂ MW: 215.05 g/mol Boiling Pt: 316 °C (1013 hPa) Melting Pt: 223…226 °C |
MDL Number:
MFCD00002567 CAS Number: 6232-88-8 EINECS: 228-343-3 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...