α-Bromo-m-toluonitrile, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-(Bromomethyl)benzonitrile, m-(Bromomethyl)benzonitrile
M-Clarity™ quality level = MQ100
145610-10G
145610-50G
SIAL145610-10GEA
180
GBP
SIAL145610-10G
SIAL145610-50G
α-Bromo-m-toluonitrile, Sigma-Aldrich®
α-Bromo-m-toluonitrile
Formula:
C₈H₆BrN MW: 196.05 g/mol Boiling Pt: 130 °C (4 mmHg) Melting Pt: 90…93 °C |
MDL Number:
MFCD00001809 CAS Number: 28188-41-2 EINECS: 248-890-1 UN: 2923 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...