α-Bromo-o-toluonitrile, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Cyanobenzyl bromide, 2-(Bromomethyl)benzonitrile, o-(Bromomethyl)benzonitrile

M-Clarity™ quality level = MQ100

144711-100G 144711-25G 144711-5G
SIAL144711-100GEA 412 GBP
SIAL144711-100G SIAL144711-25G SIAL144711-5G
α-Bromo-o-toluonitrile, Sigma-Aldrich®
α-Bromo-o-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 152…155 °C (15 mmHg)
Melting Pt: 72…74 °C
MDL Number: MFCD00001794
CAS Number: 22115-41-9
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR