p-Toluonitrile, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Methylbenzonitrile, p-Methylbenzonitrile
M-Clarity™ quality level = MQ200
132330-100G
132330-25G
132330-500G
SIAL132330-100GEA
76.9
GBP
SIAL132330-100G
SIAL132330-25G
SIAL132330-500G
p-Toluonitrile, Sigma-Aldrich®
p-Toluonitrile
Formula:
C₈H₇N MW: 117.15 g/mol Boiling Pt: 217.6 °C (1013 hPa) Melting Pt: 25…28 °C Density: 0.98 g/cm³ (20 °C) |
MDL Number:
MFCD00001827 CAS Number: 104-85-8 EINECS: 203-244-8 Merck Index: 12,09676 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...