N,O-Bis(trimethylsilyl)acetamide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate
M-Clarity™ quality level = MQ100
128910-100ML
128910-10ML
128910-1L
128910-25ML
SIAL128910-100MLEA
148
GBP
SIAL128910-100ML
SIAL128910-10ML
SIAL128910-1L
SIAL128910-25ML
N,O-Bis(trimethylsilyl)acetamide, Sigma-Aldrich®
N,O-Bis(trimethylsilyl)acetamide
Formula:
C₈H₂₁NOSi₂ MW: 203.43 g/mol Boiling Pt: 162 °C (1013 hPa) Melting Pt: ∼24 °C Density: 0.82 g/cm³ (20 °C) Flash Pt: 40 °C |
MDL Number:
MFCD00008270 CAS Number: 10416-59-8 EINECS: 233-892-7 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...