N-Ethylbenzylamine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
N-Benzylethylamine, N-Benzylethanamine
M-Clarity™ quality level = MQ100
126993-100ML
126993-25ML
SIAL126993-100MLEA
139
GBP
SIAL126993-100ML
SIAL126993-25ML
N-Ethylbenzylamine, Sigma-Aldrich®
N-Ethylbenzylamine
Formula:
C₆H₅CH₂NHCH₂CH₃ Boiling Pt: 168 °C (1013 hPa) Density: 0.909 g/cm³ (20 °C) |
MDL Number:
MFCD00009031 CAS Number: 14321-27-8 EINECS: 238-265-1 UN: 2735 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...