Methyl-10-undecenoate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Undecenylic acid methyl ester, 10-Undecenoic acid methyl ester, Methyl undec-10-enoate, Undec-10-enoic acid emthyl ester

M-Clarity™ quality level = MQ100

115126-25ML 115126-500ML
SIAL115126-25MLEA 61.9 GBP
SIAL115126-25ML SIAL115126-500ML
Methyl-10-undecenoate, Sigma-Aldrich®
Methyl 10-undecenoate
Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: –27.5 °C
Density: 0.889 g/cm³ (20 °C)
MDL Number: MFCD00016689
CAS Number: 111-81-9
EINECS: 203-910-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR