2-Methylbenzothiazole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

112143-100G 112143-25G
SIAL112143-100GEA 124 GBP
SIAL112143-100G SIAL112143-25G
2-Methylbenzothiazole, Sigma-Aldrich®
2-Methylbenzothiazole
Formula: C₈H₇NS
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: 12 °C
Density: 1.173 g/cm³ (20 °C)
MDL Number: MFCD00005794
CAS Number: 120-75-2
EINECS: 204-423-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR