(4-tert-Butylphenoxy)acetic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(4-(tert-Butyl)phenoxy)acetic acid

M-Clarity™ quality level = MQ200

0710000-25G 0710000-5G
SIAL0710000-25GEA 83.5 GBP
SIAL0710000-25G SIAL0710000-5G
(4-tert-Butylphenoxy)acetic acid, Sigma-Aldrich®
(4-tert-Butylphenoxy)acetic acid
Formula: (C₆H₄(C(CH₃)₃))CH₂COOH
MW: 208.26 g/mol
MDL Number: MFCD00021758
CAS Number: 1798-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR