1-Butyl-3-methylimidazolium chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium chloride

M-Clarity™ quality level = MQ100

04129-25G-F 04129-5G-F
SIAL04129-25G-FEA 273 GBP
SIAL04129-25G-F SIAL04129-5G-F
1-Butyl-3-methylimidazolium chloride, Sigma-Aldrich®
1-Butyl-3-methylimidazolium chloride
Formula: C₈H₁₅ClN₂
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: ∼60 °C
Density: ∼1 g/cm³ (20 °C)
Flash Pt: >110 °C
MDL Number: MFCD03095425
CAS Number: 79917-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR