5-Fluorouracil

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 5-Fluoropyrimidine-2,4(1H,3H)-dioneFluorouracil5-FU

SERA21670.04EA 0 GBP
SERA21670.04 SERA21670.02
5-Fluorouracil
5-Fluorouracil
Formula: C₄H₃FN₂O₂
MW: 130.08 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 282…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00006018
CAS Number: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1,III
Merck Index: 12,04219

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR