2,5-Diphenyloxazole

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: PPO, 2,5-Diphenyl-1,3-oxazole

3689.1 3689.2 3689.3
ROTH3689.1EA 0 GBP
ROTH3689.1 ROTH3689.2 ROTH3689.3
2,5-Diphenyloxazole
2,5-Diphenyloxazole
Formula: C₁₅H₁₁NO
MW: 221.26 g/mol
Boiling Pt: 360
Melting Pt: 70…72 °C
Density: 1.094 g/cm³ (99 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005306
CAS Number: 92-71-7
EINECS: 202-181-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR