2-Fluoro-1-iodo-4-nitrobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

035088-10G 035088-20G
FLUO035088-10GEA 0 GBP
FLUO035088-10G FLUO035088-20G
2-Fluoro-1-iodo-4-nitrobenzene
2-Fluoro-1-iodo-4-nitrobenzene
Formula: C₆H₃FINO₂
MDL Number: MFCD00033987
CAS Number: 2996-30-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR