5-Bromo-2-chloropyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
024509-25G
024509-5G
FLUO024509-25GEA
0
GBP
FLUO024509-25G
FLUO024509-5G
5-Bromo-2-chloropyridine
5-Bromo-2-chloropyridine
Formula:
C₅H₃BrClN Boiling Pt: 205 °C (1013 hPa) Melting Pt: 71 °C |
MDL Number:
MFCD01318951 CAS Number: 53939-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...