3-Bromobenzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: m-Bromobenzoyl chloride, 3-Bromobenzoylchloride

024397-10G 024397-50G
FLUO024397-10GEA 0 GBP
FLUO024397-10G FLUO024397-50G
3-Bromobenzoyl chloride
3-Bromobenzoyl chloride
Formula: C₇H₄BrClO
Boiling Pt: 74.5…75 °C (0.5 mmHg)
Density: 1.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000669
CAS Number: 1711-09-7
EINECS: 216-978-9
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR