4-Bromobenzyl mercaptan

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024343-1G 024343-5G
FLUO024343-1GEA 0 GBP
FLUO024343-1G FLUO024343-5G
4-Bromobenzyl mercaptan
4-Bromobenzyl mercaptan
Formula: C₇H₇BrS
MDL Number: MFCD03701566
CAS Number: 19552-10-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR