3-Fluorobenzyl mercaptan

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024342-1G 024342-5G
FLUO024342-1GEA 0 GBP
FLUO024342-1G FLUO024342-5G
3-Fluorobenzyl mercaptan
3-Fluorobenzyl mercaptan
Formula: C₇H₇FS
MDL Number: MFCD06798004
CAS Number: 40096-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR