2-Fluorobenzyl mercaptan

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024341-1G 024341-5G
FLUO024341-1GEA 0 GBP
FLUO024341-1G FLUO024341-5G
2-Fluorobenzyl mercaptan
2-Fluorobenzyl mercaptan
Formula: C₇H₇FS
MDL Number: MFCD00041467
CAS Number: 72364-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR